Learn More
Simport™ Scientific M480 Disposable Embedding Cassettes in Quickload™ Stacks
M480 Disposable Embedding Cassettes in Quickload™ Stacks for holding and identifying tissue samples in processing/embedding procedures.
$26.26 - $642.00
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
22-038-034
|
Simport™ Scientific
M4802T |
White |
Case of 2000 for $637.00
|
|
|||||
22-038-035
|
Simport™ Scientific
M4803T |
Pink |
Case of 2000 for $637.00
|
|
|||||
22-038-036
|
Simport™ Scientific
M4804T |
Green |
Case of 2000
for $26.26
Save 86% Reg : |
|
|||||
22-038-037
|
Simport™ Scientific
M4805T |
Yellow |
Case of 2000
for $26.26
Save 96% Reg : |
|
|||||
22-038-038
|
Simport™ Scientific
M4806T |
Blue |
Case of 2000 for $637.00
|
|
|||||
22-038-039
|
Simport™ Scientific
M4807T |
Peach |
Case of 2000 for $637.00
|
|
|||||
22-038-040
|
Simport™ Scientific
M4808T |
Tan |
Case of 2000 for $642.00
|
|
|||||
22-038-041
|
Simport™ Scientific
M4809T |
Gray |
Case of 2000 for $637.00
|
|
|||||
22-038-042
|
Simport™ Scientific
M48010T |
Lilac |
Case of 2000
for $26.26
Save 86% Reg : |
|
|||||
22-038-043
|
Simport™ Scientific
M48011T |
Orange |
Case of 2000
for $26.26
Save 86% Reg : |
|
|||||
22-038-044
|
Simport™ Scientific
M48012T |
Aqua |
Case of 2000 for $637.00
|
|
|||||
Description
- In Quickload™ stacks
- 45° Anterior writing area
- Accept standard metal lids (cat# M481)
- Suitable for automated printers
- Made of acetal
- Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes.
Certifications
CE-IVD
Specifications
2000/Pk. | |
Acetal |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.