Learn More
Lonza™ BioWhittaker™ X-VIVO™ 10 Hematopoietic Serum-Free Culture Media
For adoptive immunotherapy and cellular therapy research
Specifications
Content And Storage 2° to 8°C
Product Type Stem Cell Serum Free Media (SFM)
View More Specs
Products 1
Catalog Number Mfr. No. dimWithoutAdditives Quantity Price Quantity & Availability  
Catalog Number Mfr. No. dimWithoutAdditives Quantity Price Quantity & Availability  
BW04-380Q
View Documents
Lonza™
04380Q
1L Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area. N/A
Description
Description
  • Does not contain any lymphokines required for activation of human lymphocytes
  • Intended for in vitro procedures
  • Device Master Records (DMRs) for these products are held in an FDA Master File
  • Chemically defined media optimized for specific cell types
  • For further manufacturing or laboratory use
  • Designed to support the generation of lymphokine activated killer (LAK) cells in a serum-free environment
  • Limited by Federal (USA) law to investigational use
  • These products are for research use only
  • Not approved for human or veterinary use, for application to humans or animals, or for use in clinical or in vitro procedures
Specifications
Specifications
2° to 8°C
Stem Cell Serum Free Media (SFM)