Learn More
Lonza™ BioWhittaker™ X-VIVO™ 10 Hematopoietic Serum-Free Culture Media
For adoptive immunotherapy and cellular therapy research
Supplier: Lonza™ 04380Q
Description
- Does not contain any lymphokines required for activation of human lymphocytes
- Intended for in vitro procedures
- Device Master Records (DMRs) for these products are held in an FDA Master File
- Chemically defined media optimized for specific cell types
- For further manufacturing or laboratory use
- Designed to support the generation of lymphokine activated killer (LAK) cells in a serum-free environment
- Limited by Federal (USA) law to investigational use
- These products are for research use only
- Not approved for human or veterinary use, for application to humans or animals, or for use in clinical or in vitro procedures
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
2° to 8°C | |
Stem Cell Serum Free Media (SFM) | |
Phenol Red | |
1L |