Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LiChropur™ Hydrogen Chloride Solution, 3 M in Methanol, For GC Derivatization, MilliporeSigma™ Supelco™
Specifications
CAS | 7647-01-0 |
---|---|
Linear Formula | HCl |
MDL Number | MFCD00011324 |
Density | 0.871 to 0.881 g/mL (at 20°C) |
Synonym | Methanolic HCl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
11-102-0141
|
MilliporeSigma Supelco
9096410X1ML |
10 x 1 mL |
Each for $80.90
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
11-101-9441
|
MilliporeSigma Supelco
90964100ML |
100 mL |
Each for $116.20
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
11-101-8279
|
MilliporeSigma Supelco
90964500ML |
500 mL |
Each for $403.50
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Methanolic HCl (hydrogen chloride in methanol) is particularly useful for preparing methyl esters of volatile (short chain) fatty acids. Fatty acids are esterified by heating them with an anhydrous alcohol (e.g., methanol) in the presence of an acidic catalyst (e.g., HCl) in a sealed vessel at a high temperature for a short time. In the reaction, a fatty acid molecule and an alcohol molecule are joined, with the release of a water molecule. The derivatives can be quickly and easily recovered, quantitatively, from the reaction medium.Specifications
7647-01-0 | |
MFCD00011324 | |
Methanolic HCl |
HCl | |
0.871 to 0.881 g/mL (at 20°C) | |
GC Derivatization |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title