Learn More
LiChropur™ Hydrogen Chloride Solution, 3 M in Methanol, For GC Derivatization, MilliporeSigma™ Supelco™
Supplier: MilliporeSigma Supelco 90964500ML
Description
Methanolic HCl (hydrogen chloride in methanol) is particularly useful for preparing methyl esters of volatile (short chain) fatty acids. Fatty acids are esterified by heating them with an anhydrous alcohol (e.g., methanol) in the presence of an acidic catalyst (e.g., HCl) in a sealed vessel at a high temperature for a short time. In the reaction, a fatty acid molecule and an alcohol molecule are joined, with the release of a water molecule. The derivatives can be quickly and easily recovered, quantitatively, from the reaction medium.Specifications
7647-01-0 | |
MFCD00011324 | |
Methanolic HCl | |
GC Derivatization |
HCl | |
0.871 to 0.881 g/mL (at 20°C) | |
36.46 | |
500 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany