Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cenmed™ Latex-free Tourniquets
SDP

Cenmed™ Latex-free Tourniquets are proprietary blend of materials that gives performance of latex but is completely latex-free.

Supplier:  Cenmed™ 970001/Y

 View more versions of this product

Catalog No. 21-520-131


Add to Cart
This item is not returnable. View return policy

Description

Description

  • Completely latex-free
  • Material: Nitrile rubber
  • Thickness: 6.35mm (0.25 in.)
Specifications

Specifications

Yellow
Nitrile Rubber
0.025 in.
Yes
Tourniquet
45.7 x 2.54 cm (18 x 1 in.)
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit