Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LaMotte™ Reagent Refill
This Reagent Refill is a replacement reagent.
Supplier: LaMotte™ 4499L
Description
Available in multiple sizes:
- 500 mL bottle provides multiple tests.
- Kit size bottle.
Specifications
Reagent Refill | |
Bottle | |
Chlorine Reagent 2 |
Safety and Handling
ShelfLife : 2 Years
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction