Learn More
Mirus Bio™ Label IT™ Nucleic Acid Labeling Kit, TM-Rhodamine
Label IT™ Nucleic Acid Labeling Reagents for efficient, direct, non-enzymatic labeling of DNA and RNA.
Supplier: Mirus Bio™ MIR 4125
This item is not returnable.
View return policy
Description
- Label Any DNA or RNA Template – Suitable for a wide range of applications
- One-step Chemical Method – Easily and consistently control the labeling reactions
- Adjustable Labeling Density – Achieve high sensitivity with optimally labeled DNA and RNA
- Covalent Mechanism – Permanent, non-destructive modification of nucleic acid residues is ideal for many diverse applications; labels do not impact hybridization performance
NOTE: All the Label IT Reagents use the same technology, but each attach different fluorophores with distinct excitation and emission spectra.
Specifications
Fluorescence | |
DNA (Labeled), RNA (Labeled) | |
DNA/RNA | |
25 μg |
Alexa Fluor 546 | |
DNA/RNA | |
Label Reagent | |
Lyophilized |