Learn More
DWK Life Sciences Kimble™ Kontes™ Self Washing with Square Joint Bead Rotary Evaporator Trap
This trap is designed to eliminate hold-up common to the round style of rotary evaporator traps. Available in 100mL or 250mL capacities, with 14/20 or 24/40 bottom taper joints and 24/40 top taper joints.
Supplier: DWK Life Sciences 5703000114
Description
- Tapered sidewalls permit continuous washing with condensed solvent while preventing vapor cooling points along the path of the condenser
- The upper stem has 4 large holes that allow solvent vapor to flow freely and help to make this portion of the trap easy to clean
- The increased volume of the trap prevents “bumping” and keeps solvent out of the vapor tube
- Square Joint Bead and Standard Taper joints
- Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Upper: 24/40, Lower: 14/20 | |
3.4 oz., 100 mL | |
Borosilicate Glass | |
USP Type I and ASTM E438, Class A |
100 mL | |
Rotary Evaporator Trap | |
3.4 oz. | |
Designed to eliminate hold-up common to the round style of rotary evaporator traps |