Learn More
DWK Life Sciences Kimble™ ValueWare™ Culture Tubes With Marking Spots
With fire-polished top rim and uniform wall for maximum heat transfer and chemical resistance.
Supplier: DWK Life Sciences 5504816150
Description
- KIMAX™ KG-33 borosilicate glass
- Durable white ceramic enamel marking spot
- Reusable
Compliance
Designed from ASTM™ Specification E982, Type IV requirements
Specifications
ASTM E982, Type IV | |
150 mm | |
Reusable | |
16 mm |
24 mL | |
Borosilicate Glass | |
White Ceramic Enamel Marking Spot |