Learn More
DWK Life Sciences Kimble™ Kontes™ 2000 mL Four Vertical Neck Round Bottom Flask
SDP
Four-neck heavy wall round bottom flask with standard taper outer joints, all vertical
Supplier:  DWK Life Sciences 6070001129
 View more versions of this product
Catalog No. 12-141-477
Add to Cart
Description
Description
  • Manufactured from 33 expansion, low extractable borosilicate glass
  • Conforming to USP Type I and ASTM E438, Type I, Class A requirements
Specifications
Specifications
USP Type I, ASTM E438, Type I, Class A
Four vertical neck round bottom flask
29/42 mm
2000 mL
29/42 mm
161 mm
Product Suggestions
Product Suggestions