Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Kimble™ Brand Rotary Evaporator: Bottom Vapor Tube Traps
Accessory for rotary evaporator trap
Supplier: DWK Life Sciences 5702605024
Description
- Square Joint Bead
Specifications
Upper: 24/40, Lower: 24/40 | |
22 cm | |
Bottom Vapor Tube Trap | |
Borosilicate Glass | |
USP Type I and ASTM E438, Class A |
500 mL | |
16.9 oz., 500 mL | |
16.9 oz. | |
Designed for use between flask and rotary evaporator to reduce carry-over from foaming or bumping | |
8.6 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction