Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ Inserts for 1.5 mL Large Opening Vials

Allow the use of smaller sample volumes with standard autosampler vials.

Supplier:  MilliporeSigma™ Supelco™ 854110

 View more versions of this product

Catalog No. 11-100-6743

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

  • Vial inserts for 1.5 mL large opening vials [0.1 mL, clear glass conical (with bottom spring)] allow the use of smaller sample volumes with standard autosampler vials.
  • Inserts are generally chosen based on the inner diameter of the vial neck.
  • Large opening vials (6 mm ID) require a 6 mm OD insert.
  • Clear glass conical inserts used with CTC LC PAL autosamplers.
Specifications

Specifications

Conical Clear Glass (with Bottom Spring)
0.1 mL
1000
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit