Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Standard Output Lasers
The standard in He-Ne lasers.
Specifications
Wavelength | 632.8 nm |
---|---|
Classification | CDRF Class II |
Temperature (Metric) Operating | 0°C to 50°C |
Temperature (English) Operating | 32°F to 122°F |
Dimensions (L x W x H) | 23.9 x 7.2 x 7.4cm (9.4 x 2.8 x 2.9 in.) |
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
S42714ND
|
Industrial Fiber Optics Inc
ML810 |
He-Ne Lab Laser, 0.8 mW | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
- 0.8mW Laser is popular laser for educational use
- Output power of 0.8mW is ideal for most users in average-sized classroom or student laboratory
- Beam is visible in bright or semi-darkened room
- Dark room is suggested when it is necessary to see fine details in diffraction pattern, to make precise intensity measurements of the laser output or to make holograms
- The 0.5mW Laser is ideal for individual or group work in small rooms
Compliance
Both are Class II lasers and comply with the Laser Performance Standard of the U.S. Department of Health, Education and Welfare, U.S. Center for Devices and Radiological Health.
Specifications
632.8 nm | |
0°C to 50°C | |
23.9 x 7.2 x 7.4cm (9.4 x 2.8 x 2.9 in.) |
CDRF Class II | |
32°F to 122°F |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title