Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Standard Output Lasers
The standard in He-Ne lasers.
Supplier: Industrial Fiber Optics Inc ML810
Description
- 0.8mW Laser is popular laser for educational use
- Output power of 0.8mW is ideal for most users in average-sized classroom or student laboratory
- Beam is visible in bright or semi-darkened room
- Dark room is suggested when it is necessary to see fine details in diffraction pattern, to make precise intensity measurements of the laser output or to make holograms
- The 0.5mW Laser is ideal for individual or group work in small rooms
Compliance
Both are Class II lasers and comply with the Laser Performance Standard of the U.S. Department of Health, Education and Welfare, U.S. Center for Devices and Radiological Health.
Specifications
He-Ne Lab Laser, 0.8 mW | |
CDRF Class II | |
32°F to 122°F |
632.8 nm | |
0°C to 50°C | |
23.9 x 7.2 x 7.4cm (9.4 x 2.8 x 2.9 in.) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction