Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva HyClone™ Newborn Bovine Calf Serum, New Zealand Origin
A cost-effective alternative to Fetal Bovine Serum (FBS).
Supplier: Cytiva SH30401.02IR3045
Description
Specifications
Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Total Protein,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Osmolality,Iron,TIBC,Percent Iron Saturation,Glucose,IgG | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C | |
Sterile | |
Irradiated, 30 to 45 doses | |
≤ 25 EU/mL | |
Yes |
USDA Regulations | |
1000 mL | |
0.1 μm filtration | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
No | |
Yes |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction