Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva HyClone™ Newborn Bovine Calf Serum, New Zealand Origin
A cost-effective alternative to Fetal Bovine Serum (FBS).
$131.70 - $232.50
Specifications
Certifications/Compliance | USDA Regulations |
---|---|
Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C |
Sterility | Sterile |
Endotoxin Concentration | ≤ 25 EU/mL |
Mycoplasma Detected | Yes |
Catalog Number | Mfr. No. | Processing | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Processing | Quantity | Price | Quantity & Availability | ||||
SH304010140
|
Cytiva
SH30401.01IR2540 |
Irradiated, 25 to 40 doses | 500 mL |
Each for $133.80
|
|
||||
SH3040102HI
|
Cytiva
SH30401.02HI |
Heat inactivated | 1000 mL |
Each for $232.50
|
|
||||
SH304010240
|
Cytiva
SH30401.02IR2540 |
Irradiated, 25 to 40 doses | 1000 mL |
Each for $224.00
|
|
||||
SH3040101HI
|
Cytiva
SH30401.01HI |
Heat Inactivated | 500 mL |
Each for $131.70
|
|
||||
SH304010245
|
Cytiva
SH30401.02IR3045 |
Irradiated, 30 to 45 doses | 1000 mL |
Each for $232.50
|
|
||||
Description
Specifications
USDA Regulations | |
Sterile | |
Yes |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C | |
≤ 25 EU/mL | |
Yes |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title