Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva HyClone™ Iron-Supplemented Newborn Bovine Calf Serum (New Zealand)
A cost-effective alternative to Fetal Bovine Serum (FBS).
Supplier: Cytiva SH30626.02
Description
Specifications
Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Saturation, Glucose, IgG | |
New Zealand | |
500 mL | |
Sequential-filtered, 100 nm | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
No | |
HyClone™ |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. | |
Newborn Calf Bovine Serum | |
Sterile | |
None | |
≤ 25 EU/mL | |
≤ 25 mg/dL | |
Yes |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction