Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva HyClone™ Iron-Supplemented Newborn Bovine Calf Serum (New Zealand)
A cost-effective alternative to Fetal Bovine Serum (FBS).
$245.44 - $372.00
Specifications
Additional Assays | Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Saturation, Glucose, IgG |
---|---|
Content And Storage | Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. |
Country of Origin | New Zealand |
Sterility | Sterile |
Sterilization Method | Sequential-filtered, 100 nm |
Catalog Number | Mfr. No. | Quantity | Processing | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Processing | Price | Quantity & Availability | ||||
SH3062603
|
Cytiva
SH30626.03 |
1000 mL | None |
Each for $372.00
|
|
||||
SH3062603HI
|
Cytiva
SH30626.03HI |
1000 mL | Heat Inactivated |
Each for $245.44
|
|
||||
Description
Specifications
Gamma Globulin, Alkaline Phosphatase, Lactate Dehydrogenase, SGPT, SGOT, pH, Total Protein, Albumin, Blood Urea Nitrogen, Creatinine, Total Bilirubin, Sodium, Potassium, Calcium, Chloride, Inorganic Phosphorus, Osmolality, Iron, TIBC, Percent Saturation, Glucose, IgG | |
New Zealand | |
Sequential-filtered, 100 nm | |
HyClone™ |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. | |
Sterile | |
≤ 25 mg/dL | |
Yes |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title