Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Medicom™ Hopen™ SafeFeet™ SkidGuard™ Shoe Covers
Medicom™ Hopen™ SafeFeet™ SkidGuard™ Shoe Covers with comfortable, microporous soles offer optimal protection
Supplier: Medicom™ 13.007/05
This item is not returnable.
View return policy
Description
- Polypropylene (PP) shoecovers with Polyethylene (PE) plastic-coated sole prevent users from slipping and ensures a long product life
- Shoe covers have the ability to face the risk of abrasion on rough floors without tearing
- Adaptable to the different morphologies of all types of shoes
- Available in multiple colors, providing the ability of color code tracking
- Due to its large size and height, this model fits perfectly to all types of safety shoes
-
Specifications
42 cm x 230 cm | |
Polyethylene | |
Single-use | |
Non-skid | |
Abrasion |
Polypropylene | |
White | |
Non-sterile | |
Dispenser Box | |
Shoe Covers |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction