Learn More
Medicom™ Hopen™ SafeFeet™ SkidGuard™ Shoe Covers
Medicom™ Hopen™ SafeFeet™ SkidGuard™ Shoe Covers with comfortable, microporous soles offer optimal protection
Supplier: Medicom™ 12.007/05
This item is not returnable.
View return policy
Description
- Polypropylene (PP) shoecovers with Polyethylene (PE) plastic-coated sole prevent users from slipping and ensures a long product life
- Shoe covers have the ability to face the risk of abrasion on rough floors without tearing
- Adaptable to the different morphologies of all types of shoes
- Available in multiple colors, providing the ability of color code tracking
- Due to its large size and height, this model fits perfectly to all types of safety shoes
-
Specifications
2X-Large | |
Polyethylene | |
Single-use | |
Non-skid | |
Abrasion |
Polypropylene | |
White | |
Non-sterile | |
Bagged | |
Shoe Covers |