Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Heavy Duty HDPE Lab Trays
SDPSCI_ED

Fabricated with 1/4 in. thich high density polyethylene.

 View more versions of this product

Catalog No. S37152


Add to Cart

Description

Description

  • Rectangular with welded corners and bottom
  • Can be customized with spigots and covers
  • Won't rust

Order Info

Additional spill containment tray sizes available. To order, contact your Fisher Scientific Customer Service Representative.

Specifications

Specifications

16 x 16 x 8 in. (40.5 x 40.5 x 20.32 cm)
40.5 cm
Laboratory Tray
Welded corners and bottom
8 in.
0.48 cm
Non-Autoclavable
20.32 cm
HDPE
40.5 cm
16 in.
Rectangular
16 in.
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit