Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hamilton™ Modified Microliter™ Syringe, Special Termination, Sold by MilliporeSigma™ Supelco™
Sample is contained in the needle
Supplier: MilliporeSigma Supelco 26217
Specifications
Autoclavable | |
26s G | |
0.13 mm | |
0.47 mm | |
Microliter™ Syringe | |
239°F | |
50°F | |
1 | |
1 Ea. | |
Hamilton | |
Cone | |
Special |
Borosilicate Glass (Barrel), Stainless Steel (Tip) | |
0.005 in. | |
0.0187 in. | |
1.71 in. | |
6000 psig (413.7 bar) | |
115°C | |
7001 Special Agilent | |
10°C | |
1 μL | |
43 mm | |
Agilent 7673, 7683, 7693A and 6850 Autosamplers |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Microliter is a trademark of Hamilton Co.
Spot an opportunity for improvement?
Provide Content Correction