Learn More
Hamilton™ 1000 Series Syringe, Fixed Needle with Side Port, Sold by MilliporeSigma™ Supelco™
Gas-tight design minimizes gas exchange during GC analysis. Syringes provide accurate volume delivery with a plunger that is mostly unsurpassed.
Supplier: MilliporeSigma Supelco 20710U
Specifications
Not Autoclavable | |
22 G | |
0.41 mm | |
0.72 mm | |
Gastight™ Syringe | |
122°F | |
50°F | |
5 | |
1 Ea. | |
Hamilton | |
Cone with Side Port |
Borosilicate Glass (Barrel), PTFE (Plunger), Stainless Steel (Tip) | |
0.0161 in. | |
0.0283 in. | |
2 in. | |
200 psig (13.8 bar) | |
50°C | |
1002 LTN | |
10°C | |
2.5 mL | |
51 mm | |
Luer Tip, Cemented |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
GASTIGHT is a registered trademark of Hamilton Co.