Learn More
Promega 5X Colorless GoTaq™ Reaction Buffer
The 5X Green GoTaq Reaction Buffer contains a compound that increases sample density as well as two dyes that separate during electrophoresis to show migration progress. Samples can be loaded directly onto gels without the need for loading dye.
Supplier:  Promega M7921
Catalog No. PRM7921
Add to Cart
Description
Description
  • Buffers for Reliable, Robust PCR
  • Available with green dye for direct-to-gel convenience, or in colorless format for applications where absorbance or fluorescence measurements are necessary prior to PCR cleanup
  • Contain 7.5mM MgCl2 for a final concentration of 1.5mM in a 1X reaction
Specifications
Specifications
−30 to −10°C
20mL
Product Suggestions
Product Suggestions