Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycerol, ACS Reagent, for organic synthesis, prep-LC, and general laboratory use, >99.0%, Honeywell
Click to view available options
Quantity:
4 x 4 L
Packaging:
Poly Bottle
Specifications
CAS Min % | 99.7000% |
CAS Max % | 100.0000% |
Density | 1.261 g/cm3 |
Boiling Point | 290°C |
Flash Point | 199°C |
Loss on Drying | 0.01% max. |
Packaging | Poly Bottle |
Quantity | 4 x 4 L |
Formula Weight | 92.1 |
Percent Purity | >99.7% |
Show More |
Safety and Handling
ShelfLife | 547 days from date of manufacture |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction