Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Glass Volumetric Flask with White Graduations and Stopper
Volumetric Flask, Class A, ASTM. Blue Snap Cap, Single, White Graduation. Made of Borosilicate Glass.
Supplier: Eisco™ CH0443A01WTST
Description
- Made of high quality, heavy duty, Borosilicate 3.3 glass
- Single white graduation mark
- Class A tolerance. Compliant with ASTM-E288 standards
- Fitted with blue snap cap
Specifications
5 mL | |
Volumetric Flask | |
Reusable | |
Beaded | |
Class A | |
Uncoated |
Borosilicate Glass (3.3) | |
Autoclavable | |
No | |
Non-sterile | |
Flat |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction