Learn More
Eisco™ Glass Buchner Filtering Flasks
High Quality borosilicate 3.3 glass Buchner filtering flasks available in a variety of socket sizes
Supplier: Eisco™ CH0422D01
Description
Warranty and Services
1 year limited manufacturer warranty
Specifications
Clear | |
Borosilicate Glass (3.3) | |
29/32 | |
Yes |
500 mL | |
Filtering Liquids | |
Autoclavable | |
Non-sterile |