Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Innovating Science™ Genetics of Blood Types Using Simulated Blood Kit
Learn how genetics determine blood type and possible inheritance patterns.
$29.50 - $56.75
Specifications
Certifications/Compliance | 49 CFR 173.4 |
---|---|
Color | White |
Grade | 7 to 14 |
Age | 13+ years |
For Use With (Application) | For fictional paternity test |
Includes:
- Four simulated blood samples
- One bottle each anti-A and anti-B serum
- 40 blood typing trays
Description
Students use simulated blood to determine the blood type of four unknown samples and then use their results to assist in the resolution of a fictional paternity dispute.
- Provides the most accurate simulation of the actual blood typing procedure
- No toothpicks, stirring or waiting for results
- Hands-on activities
- Kit contains materials for 10 groups
Uses only simulated blood. These activities do not use real blood.
Specifications
49 CFR 173.4 | |
7 to 14 | |
For fictional paternity test | |
1 |
White | |
13+ years | |
4 simulated blood samples, mother sample x, child sample y, 1 bottle simulated anti-a serum, 1 bottle simulated anti-b serum, 40 blood typing trays, Teacher's manual and student study guide copymasters | |
10 Lab Groups |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title