Learn More
Innovating Science™ Genetics of Blood Types Using Simulated Blood Kit
Learn how genetics determine blood type and possible inheritance patterns.
Supplier: Innovating Science™ IS3102REF
Includes:
- Four simulated blood samples
- One bottle each anti-A and anti-B serum
- 40 blood typing trays
Description
Students use simulated blood to determine the blood type of four unknown samples and then use their results to assist in the resolution of a fictional paternity dispute.
- Provides the most accurate simulation of the actual blood typing procedure
- No toothpicks, stirring or waiting for results
- Hands-on activities
- Kit contains materials for 10 groups
Uses only simulated blood. These activities do not use real blood.
Specifications
49 CFR 173.4 | |
White | |
13+ years | |
4 simulated blood samples, mother sample x, child sample y, 1 bottle simulated anti-a serum, 1 bottle simulated anti-b serum, 40 blood typing trays, Teacher's manual and student study guide copymasters | |
10 Lab Groups |
Blood Typing Genetics Refill | |
7 to 14 | |
For fictional paternity test | |
1 |