Learn More
Thermo Scientific™ GeneJET PCR Purification Kit PROMO
GeneJET PCR Purification Kit utilizes a proprietary silica-based membrane technology in the form of a convenient spin column, eliminating the need for tedious resin manipulations or toxic phenol-chloroform extractions.
$69.94 - $308.14
Specifications
Format | Spin Column |
---|---|
High-throughput Compatibility | Not High-throughput Compatible (Manual) |
Run Time | 5 min. |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FERK0701PM
|
Thermo Scientific™
K0701 |
50 Preps |
Each for $69.94
|
|
|||||
FERK0702PM
|
Thermo Scientific™
K0702 |
250 Preps |
Each for $308.14
|
|
|||||
Description
Thermo Scientific GeneJET PCR Purification Kit utilizes a proprietary silica-based membrane technology in the form of a convenient spin column, eliminating the need for tedious resin manipulations or toxic phenol-chloroform extractions. It effectively removes primers, dNTPs, unincorporated labeled nucleotides, enzymes, and salts from PCR and other reaction mixtures.
The kit can be used for purification of DNA fragments from 25 bp to 20 kb with recovery rates up to 100%. Each GeneJET purification column has a total binding capacity of up to 25 μg of DNA, and the entire procedure takes 5 minutes. The purified DNA can be used in common downstream applications, such as sequencing, restriction digestion, labeling, ligation, cloning, in vitro transcription, blotting or in situ hybridization.
Highlights
- Highly efficient: 90 to 100% recoveries in the range of 100 bp to 10 kb
- Fast: procedure takes only 5 minutes
- Convenient: spin columns are capped and assembled with collection tubes
Applications
- Fast and efficient purification of DNA fragments ideal for use in all conventional molecular biology procedures including:
- FastDigest or conventional restriction digestion
- Automated fluorescent or radioactive sequencing
- PCR
- In vitro transcription
- In situ hybridization
- Labeling
- Cloning
Specifications
Spin Column | |
Not High-throughput Compatible (Manual) | |
5 min. |
For Research Use Only. Not for use in diagnostic procedures.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.