Learn More
Fischer Technical Company™ Fixed Volume Mini Pipettors
Fixed Volume Mini Pipettors offer accuracy at an affordable price.
$28.72
Specifications
Autoclavable | Autoclavable |
---|---|
Disposable | Reusable |
For Use With | Micropipette Tip |
Material | Polypropylene |
Model | D590 |
Description
- Unique miniature size pipettors provide accurate fixed volume dispensing
- Ideal for micro-volumes and gel loading applications
- Accept universal fit disposable pipette tips
- Accepts 200 μL tips
- Available in two sizes, color-coded for easy identification
- Accuracy 0.3 μL and 0.6 μL, respectively
Specifications
Autoclavable | |
Micropipette Tip | |
D590 | |
Non-sterile |
Reusable | |
Polypropylene | |
1 | |
Fixed Volume Minipipette |