Learn More
Fischer Technical Company™ Fixed Volume Mini Pipettors
Fixed Volume Mini Pipettors offer accuracy at an affordable price.
Supplier: Fischer Technical Company™ 347021
Description
- Unique miniature size pipettors provide accurate fixed volume dispensing
- Ideal for micro-volumes and gel loading applications
- Accept universal fit disposable pipette tips
- Accepts 200 μL tips
- Available in two sizes, color-coded for easy identification
- Accuracy 0.3 μL and 0.6 μL, respectively
Specifications
Autoclavable | |
Reusable | |
Polypropylene | |
1 | |
Fixed Volume Minipipette |
Pink | |
Micropipette Tip | |
D590 | |
Non-sterile | |
20 μL |