Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Fixed Volume Micropipettes
Ideal for clinical diagnostic laboratories and routine tests. Eisco™ Fixed Volume Micropipettes are designed to deliver precise liquid volumes using air displacement technology.
Supplier: Eisco™ CH0586N
This item is not returnable.
View return policy
Description
- Fixed volume micropipette
- Full size
- 2 stop plunger for sample intake/dispense
- Air displacement technology
Specifications
S39856, S39865, S38972 | |
Black | |
Micropipette Tip | |
Non-sterile | |
1000 μL | |
Micropipette Tips | |
Mechanical tip ejector | |
30 x 8 x 4 cm |
Partial | |
Reusable | |
Plastic | |
Micropipette | |
Clinical Diagnostics, Assays | |
1000 μL | |
1 Year |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction