Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Fixed Volume Micropipettes
Ideal for clinical diagnostic laboratories and routine tests. Eisco™ Fixed Volume Micropipettes are designed to deliver precise liquid volumes using air displacement technology.
$43.00
Specifications
Autoclavable | Partial |
---|---|
Color | Black |
Disposable | Reusable |
For Use With | Micropipette Tip |
Material | Plastic |
Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Price | Quantity & Availability | |||||
S39845
|
Eisco™
CH0586F |
20 μL |
Each for $43.00
|
|
|||||
S39853
|
Eisco™
CH0586N |
1000 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Fixed volume micropipette
- Full size
- 2 stop plunger for sample intake/dispense
- Air displacement technology
Specifications
Partial | |
Reusable | |
Plastic | |
Micropipette | |
Micropipette Tips |
Black | |
Micropipette Tip | |
Non-sterile | |
Clinical Diagnostics, Assays | |
1 Year |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title