Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
First Safety™ Gas Cylinder Chain Set
Welded chain set includes bit snap for easy chain connection
Supplier: First Safety™ 35366
Description
This set can be used with First Safety wall and bench mounted cylinder brackets, cylinder stands, racks, process stations and cylinder carts.
- £1/0 heavy gauge with twisted loop to enable chain to sit flat against cylinder for minimum chance of damaging the cylinder finish
- 104.1cm (41 in.) long with included bit snap
- Color: Silver
Specifications
41 in. | |
1/0g | |
Chain Set |
104.1 cm | |
Bit snap | |
First safety wall and bench mounted cylinder brackets, cylinder stands, racks, process stations and cylinder carts |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title