Learn More
Gibco™ ExpiSf™ Expression System Starter Kits
The ExpiSf Expression System is a complete chemically-defined baculovirus-insect cell protein expression system that delivers superior yields and consistent performance run after run using a fast, streamlined workflow.
$2155.00
Specifications
Type | Insect Cell Protein Expression System |
---|---|
Delivery Type | Baculovirus |
Shipping Condition | Approved for shipment at Room Temperature or on Wet or Dry Ice |
Cell Line | Sf9, SFM Adapted |
Product Line | ExpiSf™ |
Catalog Number | Mfr. No. | Type | Format | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Format | Price | Quantity & Availability | ||||
A38841
|
Gibco™
A38841 |
Insect Cell Protein Expression System | Kit with Erlenmeyer Flasks |
Each of 1 for $2,155.00
|
|
||||
Description
The ExpiSf Expression System is a complete chemically-defined baculovirus-insect cell protein expression system that delivers superior yields and consistent performance run after run using a fast, streamlined workflow.
The ExpiSf Expression System offers
- A chemically defined, yeastolate-free, animal origin-free medium for consistent cell growth and protein expression
- Integrated components that work in concert to deliver three times higher protein yields compared to traditional insect expression platforms
- Robust production of high-titer, high-quality P0 recombinant baculovirus in suspension culture format without the need for further virus amplification
- A fast, simplified workflow that allows you to obtain protein in as little as 6–10 days; half the time of traditional protocols
- Scalable expression that can be applied to cultures from 24-deep well plates to >3 L shake flasks
Specifications
Insect Cell Protein Expression System | |
Approved for shipment at Room Temperature or on Wet or Dry Ice | |
ExpiSf™ |
Baculovirus | |
Sf9, SFM Adapted |
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.