Learn More
United Scientific™ Erlenmeyer Flask, 24/40 Joint
Made from ASTM E-438 Type-1 Class A, Borosilicate 3.3 glass
Supplier: United Scientific™ UNJTERFL500
Description
- Borosilicate glass flasks are designed to give the best combination of thermal shock resistance and physical strength.
- Feature 24/40 jointed sockets and graduations in durable white enamel.
- Made from ASTM E-438 Type-1 Class A, Borosilicate 3.3 glass.
- These flasks comes with ground joints manufactured as per ASTM E-1404, Type 2, Class-1 requirement.
- The flasks are printed in white color enamel.
Specifications
500 mL | |
Autoclavable | |
Yes | |
Non-sterile | |
24/40 Joint |
Borosilicate Glass (3.3) | |
Reusable | |
Joint | |
Flat | |
No |