Learn More
Wearwell™ ErgoDeck Diamond Plate General Purpose Solid Matting
Wearwell™ ErgoDeck with Diamond-Plate has a box-spring foundation that compresses under foot, providing anti-fatigue benefits plus the necessary structure to support heavy foot traffic
Supplier:  Wearwell™ 590.78X18X18BKCS10
 View more versions of this product
Catalog No. 19-060-520
Add to Cart
Description
Description
  • Standing relief in wet or dry applications
  • Customize in 3-inch increments to fit any area or configuration
  • Positive Interlocking System prevents separation
  • Easily replace single pieces or sections as needed to reduce costs
  • 100% Silicone Free PVC tiles provide maximum durability
  • Diamond-Plate surface pattern delivers dependable traction and offers an Industrial look
Specifications
Specifications
Oily areas
22 mm
Diamond plate pattern
7/8 in.
Black
18 x 18 in. (46 x 46 cm)
Product Suggestions
Product Suggestions