Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Terumo Engauge Safety Hypodermic Syringe with Safety Needle
Engauge Safety Hypodermic Syringe with Safety Needle features three different modes of activation: Thumb, Finger and Hard Surface.
Supplier: Terumo SENG03L2525R
Description
- Low Dead Space
- 3 modes of activation: Thumb, Finger and Hard Surface
- Economical option
Specifications
Not Autoclavable | |
25 G | |
1 in. | |
3 mL | |
Hypodermic Needle | |
Yes |
Polypropylene (Barrel), Polypropylene (Plunger), Polypropylene (Hub) | |
Yes | |
Single-use | |
Bevel | |
Luer Lock | |
Sterile |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction