Learn More
CDS Analytical™ Empore™ SDB-XC StageTips
SDB-XC (styrenedivinylbenzene) polymer sorbent StageTips are SPE micropipette tips for the desalting of large peptides and proteins, as well as peptide fractionation.
Supplier: CDS Analytical™ 70201910037
This item is not returnable.
View return policy
Description
The CDS™ Empore™ StageTips SDB-XC (styrenedivinyl benzene) polymer sorbent solid phase extraction micropipette tips have two layers for increased capacity.
- Combines good retention capacity with high recoveries
- Recommended for desalting of large peptides and proteins and peptide fractionation at acidic, neutral, and basic pH conditions
Specifications
12 μm | |
C8 (Octyl, Dibutyl) | |
960 Cs. |
Empore | |
200 μL |