Learn More
CDS Analytical™ Empore™ SDB-XC StageTips
SDB-XC (styrenedivinylbenzene) polymer sorbent StageTips are SPE micropipette tips for the desalting of large peptides and proteins, as well as peptide fractionation.
$189.80 - $1324.00
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
13-110-059
|
CDS Analytical™
70201910037EA |
96 Pk. |
Pack of 96 for $189.80
|
|
|||||
13-110-060
|
CDS Analytical™
70201910037 |
960 Cs. |
Case of 960 for $1,324.00
|
|
|||||
Description
The CDS™ Empore™ StageTips SDB-XC (styrenedivinyl benzene) polymer sorbent solid phase extraction micropipette tips have two layers for increased capacity.
- Combines good retention capacity with high recoveries
- Recommended for desalting of large peptides and proteins and peptide fractionation at acidic, neutral, and basic pH conditions
Specifications
12 μm | |
Empore | |
200 μL |