Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDS Analytical™ Empore™ Cation StageTips
Cation exchange polymer sorbent StageTips are for peptide fractionation at acidic, neutral, and basic pH conditions.
$189.80 - $1324.00
Specifications
Particle Size | 12 μm |
---|---|
Product Line | Empore |
Sorbent | Polystyrene-divinylbenzene SCX |
Volume (Metric) | 200 μL |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
13-110-065
|
CDS Analytical™
70201910064EA |
96 Pk. |
Pack of 96 for $189.80
|
|
|||||
13-110-066
|
CDS Analytical™
70201910064 |
960 Cs. |
Case of 960 for $1,324.00
|
|
|||||
Description
The CDS™ Empore™ StageTips cation exchange (SCX) polymer sorbent solid phase extraction micropipette tips have two layers for increased capacity.
- Combines good retention capacity with high recoveries
- Recommended for peptide fractionation at acidic, neutral, and basic pH conditions
Specifications
12 μm | |
Empore | |
Polystyrene-divinylbenzene SCX | |
200 μL |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title