Learn More
CDS Analytical™ Empore™ Cation StageTips
SDP
Cation exchange polymer sorbent StageTips are for peptide fractionation at acidic, neutral, and basic pH conditions.
Supplier:  CDS Analytical™ 70201910064
 View more versions of this product
Catalog No. 13-110-066
Explore available promotions
Add to Cart
This item is not returnable. View return policy
Description
Description
The CDS™ Empore™ StageTips cation exchange (SCX) polymer sorbent solid phase extraction micropipette tips have two layers for increased capacity.
  • Combines good retention capacity with high recoveries
  • Recommended for peptide fractionation at acidic, neutral, and basic pH conditions
Specifications
Specifications
12 μm
Polystyrene-divinylbenzene SCX
960 Cs.
Empore
200 μL
Product Suggestions
Product Suggestions