Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Edvotek™ Protein InstaStain™ sheets
Stain gels faster than conventional methods avoiding large amounts of liquid stain and waste disposal.
$83.00
Specifications
Product Type | Protein Stain |
---|---|
Form | Sheet, Solid |
For Use With (Application) | To stain Protein bands on Gel after electrophoresis |
Catalog Number | Mfr. No. | Sufficient For | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Sufficient For | Price | Quantity & Availability | |||||
S94633
|
Edvotek™
2016 |
15 Gels |
Each for $83.00
|
|
|||||
S94634
|
Edvotek™
2017 |
30 Gels | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Protein InstaStain™ sheets stain gels faster than conventional methods. Protein InstaStain™ gives high quality and uniform gel staining with excellent results for photography. They are also environmentally friendly because they use a solid matrix, avoiding large amounts of liquid stain and waste disposal. Protein InstaStain can be used to stain the most common gels of various sizes (ranging from 8x8 cm to 13x13 cm)Order Info
Dimension: 10 x 10cm
Specifications
Protein Stain | |
Sheet, Solid | |
To stain Protein bands on Gel after electrophoresis |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title