Learn More
Edvotek™ Protein InstaStain™ sheets
Stain gels faster than conventional methods avoiding large amounts of liquid stain and waste disposal.
Supplier: Edvotek™ 2017
Description
- Made of environmentally safe solid matrix
- Gives high quality and uniform gel staining
- Excellent results for photography
Order Info
Dimension: 10 x 10cm
Specifications
Protein Stain | |
Sheet, Solid | |
To stain Protein bands on Gel after electrophoresis | |
30 Gels |