Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Edvotek™ PCR-Based DNA Typing
DNA typing adapted from FBI technology.
$281.00
Specifications
Quantity | 1 Kit |
---|---|
For Use With (Application) | Standard PCR |
No. of Reactions | 25 |
Requires:
- Thermal cycler
- Horizontal gel electrophoresis apparatus
- Microcentrifuge
- Automatic micropipets
- UV transilluminator
- 500mL flasks
- Glassware
- All necessary safety equipment
- Ice
- Ice bucket
- Distilled or deionized water
Description
Safe experiment for classroom use to demonstrate DNA fingerprinting.
Alu-Human DNA Typing (S66823)
- Examine students' DNA for an Alu element on Chromosome 16
- Investigate DNA patterns
- Compare Alu genotypes with those of their classmates
VNTR Human DNA Typing (S66824)
- Use PCR to fingerprint students' DNA at the D1S80 locus on Chromosome 1
- Provide an example of the variable number of tandem repeat (VNTR) polymorphism
Specifications
1 Kit | |
Standard PCR | |
25 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title