Learn More
Thermo Scientific™ DreamTaq™ Hot Start Green PCR Master Mix
Thermo Scientific™ DreamTaq™ Hot Start Green PCR Master Mix is a ready-to-use 2X reaction mix that includes DreamTaq™ Hot Start DNA Polymerase, DreamTaq Green buffer, magnesium, and dNTPs.
$213.00 - $940.00
Specifications
GC-Rich PCR Performance | Low |
---|---|
Polymerase | DreamTaq DNA Polymerase |
Reaction Speed | Standard |
Shipping Condition | Dry Ice |
For Use With (Application) | Hot-start PCR |
Description
Thermo Scientific™ DreamTaq™ Hot Start Green PCR Master Mix is a ready-to-use 2X reaction mix that includes DreamTaq™ Hot Start DNA Polymerase, DreamTaq Green buffer, magnesium, and dNTPs. The master mix allows easy reaction setup with fewer pipetting steps, with only primers, template, and water needing to be added. The green master mix includes density reagent and two tracking dyes for convenient direct loading of PCR products on a gel.
DreamTaq Hot Start DNA Polymerase is an enhanced hot start Taq DNA polymerase that enables higher PCR specificity, sensitivity, and yields compared to conventional hot start Taq DNA polymerases.
DreamTaq Hot Start DNA Polymerase employs antibody-based inhibition of DNA polymerase activity at ambient temperatures to prevent the amplification of non-specific products prior to the amplification step. With DreamTaq Hot Start DNA Polymerase, reactions can be set up at room temperature using the same protocol and cycling conditions as conventional Taq DNA polymerases.
DreamTaq Hot Start Green PCR Master Mix is formulated to enhance productivity through:
- Better reaction outcomes
- Higher yield of target amplicons from low template amounts
- Increased reaction specificity due to reliable hot-start technology
- Wider range of amplicon lengths—routine amplification of genomic DNA fragments up to 6 kb
- Enhanced convenience
- Room temperature reaction set-up
- Minimized optimization of Mg2+ concentration and of primer annealing temperatures due to optimized reaction buffer
- Simplified workflow and minimized pipetting steps with 2X master mix format
- Direct loading of PCR products on a gel
Applications
- Routine PCR
- Colony PCR
- Genotyping
- RT-PCR
- Generation of PCR products for TA cloning
Specifications
Low | |
Standard | |
Hot-start PCR | |
Built-In Hot Start | |
SuperMix or Master Mix |
DreamTaq DNA Polymerase | |
Dry Ice | |
2X | |
3'-A |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.