Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ DreamTaq™ Hot Start Green DNA Polymerase
Thermo Scientific DreamTaq Hot Start DNA Polymerase is the product of choice when you need consistently robust amplification with minimal optimization steps
$102.65 - $4152.00
Specifications
GC-Rich PCR Performance | Low |
---|---|
Polymerase | DreamTaq DNA Polymerase |
Reaction Speed | Standard |
Product Line | DreamTaq™ |
Shipping Condition | Wet or Dry Ice |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEREP1711
|
Thermo Scientific™
EP1711 |
200 U |
Each
for $102.65
Save 14% Reg : |
|
|||||
FEREP1712
|
Thermo Scientific™
EP1712 |
500 U |
Each for $270.00
|
|
|||||
FEREP1713
|
Thermo Scientific™
EP1713 |
2500 U |
Each for $1,126.00
|
|
|||||
FEREP1714
|
Thermo Scientific™
EP1714 |
4 x 2500 U |
Each for $4,152.00
|
|
|||||
Description
Thermo Scientific™ DreamTaq™ Hot Start DNA Polymerase is an enhanced hot start Taq DNA polymerase that ensures higher PCR specificity, sensitivity and yields compared to conventional hot start Taq DNA polymerases. DreamTaq Hot Start DNA Polymerase uses antibody based inhibition of DNA polymerase activity at ambient temperatures, thus preventing the amplification of non-specific products prior to the amplification step.Better reaction outcomes:
- Higher yield of target amplicons from low template amounts.
- Increased reaction specificity due to reliable hot-start technology.
- Wider range of amplicon lengths - routine amplification of genomic DNA fragments up to 6kb.
Enhanced convenience:
- Room temperature reaction set-up.
- Minimized optimization of Mg2+ concentration and of primer annealing temperatures due to optimized reaction buffer.
- Direct loading of PCR products on a gel with DreamTaq Green buffer.
Specifications
Low | |
Standard | |
Wet or Dry Ice | |
3'-A |
DreamTaq DNA Polymerase | |
DreamTaq™ | |
Built-In Hot Start | |
Separate Components |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title