Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Cover Glasses: Circles
Made of finest optical borosilicate glass, with uniform thickness and size.
This item is not returnable.
View return policy
Description
- Circular shape
- Corrosion-resistant
- Lint-free and ready to use
- Available in squares, circles and rectangles
Specifications
Coverslip | |
Borosilicate Glass | |
18 mm | |
Microscopes |
Circle | |
#1 (0.13 to 0.16 mm) | |
Circles No. 1 | |
1 oz./Pk. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction